BG Succinimidyl Ester
Lieferant: AAT BIOQUEST
Achtung
12564
AATB12564EA 292 EUR
AATB12564
BG Succinimidyl Ester
BG Succinimidyl Ester
SNAP protein labeling system enables the specific, covalent attachment of virtually any molecule to a protein of interest.
  • CLIP tag building block
  • High quality
  • Multiple applications
ADR: 5 II