BG azide
Lieferant: AAT Bioquest
12568
AATB12568EA 266 EUR
AATB12568
BG azide
BG azide
BG azide is an alkyne-reactive building block for preparing SNAP substrates from the alkyne-reactive forms of labels or biological targets.
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only