BG alkyne
Lieferant: AAT BIOQUEST
12569
AATB12569EA
266
EUR
AATB12569
BG alkyne
BG alkyne
BG alkyne is an azide-reactive building block for preparing SNAP substrates from the azide-reactive forms of labels or biological targets.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only