BG alkyne
Lieferant: AAT BIOQUEST
BG alkyne is an azide-reactive building block for preparing SNAP substrates from the azide-reactive forms of labels or biological targets.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only