Cy3 Bis NTA-Ni komplex, HIS Lite™
Lieferant: AAT BIOQUEST
Achtung
12610
AATB12610EA 614 EUR
AATB12610
Cy3 Bis NTA-Ni komplex, HIS Lite™
Cy3 Bis NTA-Ni
Polyhistidine is one of the most popular affinity tags incorporated into recombinant proteins.
  • Useful for detecting histidine-tagged proteins
  • Well-cited building block
  • Multiple applications
ADR: 5,II