HIS Lite™ Recombinant Bis NTA-Ni Complex, Cy5
Lieferant: AAT BIOQUEST
12613
AATB12613EA 614 EUR
AATB12613
HIS Lite™ Recombinant Bis NTA-Ni Complex, Cy5
Proteine und Peptide
The Cy5 Bis NTA-Ni Complex can be directly applied either to an SDS-PAGE gel or western blot membrane for fluorescence imaging.
  • Specific and highly-sensitive detection
  • Minimal crossreactivity
No secondary reaction is required since the Ni-NTA complexes are directly conjugated to the fluorophores.
Bitte beachten For research use only.