OG488-Tris NTA-Ni komplex, HIS Lite™
Lieferant: AAT BIOQUEST
|
Achtung
|
12615
AATB12615EA
307
EUR
AATB12615
OG488-Tris NTA-Ni komplex, HIS Lite™
OG488-Tris NTA-Ni
Fluorescent tris-NTA compounds provide an efficient method for site-specific and stable noncovalent fluorescence labeling of polyhistidine-tagged proteins.
- Useful for detecting histidine-tagged proteins
- Well-cited building block
- Multiple applications
ADR:
5,II |