OG488-Tris NTA-Ni komplex, HIS Lite™
Lieferant: AAT BIOQUEST
Achtung
12615
AATB12615EA 307 EUR
AATB12615
OG488-Tris NTA-Ni komplex, HIS Lite™
OG488-Tris NTA-Ni
Fluorescent tris-NTA compounds provide an efficient method for site-specific and stable noncovalent fluorescence labeling of polyhistidine-tagged proteins.
  • Useful for detecting histidine-tagged proteins
  • Well-cited building block
  • Multiple applications
ADR: 5,II