HIS Lite™ iFluor® 647 Tris NTA-Ni Complex
Lieferant: AAT BIOQUEST
12618
AATB12618EA
271
EUR
AATB12618
HIS Lite™ iFluor® 647 Tris NTA-Ni Complex
Proteinkonjugate und Markierungen
iFluor 647-Tris NTA compound is used as a sensitive fluorescent probe for detecting polyhistidine-labelled proteins in cells, solution and solid surfaces.
- Stable for 12 months
Quality tested in-house.
Bitte beachten For research use only.