HIS Lite™ Cy5 Tris NTA-Ni Complex
Lieferant: AAT BIOQUEST
12619
AATB12619EA
271
EUR
AATB12619
HIS Lite™ Cy5 Tris NTA-Ni Complex
Proteinkonjugate und Markierungen
Cy5-Tris NTA compound is used as a sensitive fluorescent probe for detecting polyhistidine-labelled proteins in cells, solution and solid surfaces.
- Stable for 12 months
Quality tested in-house.
Bitte beachten For research use only.