HIS Lite™ Cy3 Tris NTA-Ni Complex
Lieferant: AAT BIOQUEST
12620
AATB12620EA 271 EUR
AATB12620
HIS Lite™ Cy3 Tris NTA-Ni Complex
Proteinkonjugate und Markierungen
Cy3-Tris NTA compound is used as a sensitive fluorescent probe for detecting polyhistidine-labelled proteins in cells, solution and solid surfaces.
  • Stable for 12 months
Quality tested in-house.
Bitte beachten For research use only.