6XHis-DBCO Konjugat
Lieferant: AAT BIOQUEST
12623
AATB12623EA
410
EUR
AATB12623
6XHis-DBCO Konjugat
6XHis-DBCO
6XHis-DBCO conjugate is a robust building block to introduce a 6XHis-Tag into a biological molecule through the well-known click chemistry.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only