6XHis alkyne
Lieferant: AAT BIOQUEST
6XHis alkyne is an excellent building block to make 6XHis conjugates for developing His tag detection probes and purification tools through the well-known click chemistry.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only