6XHis alkyne
Lieferant: AAT BIOQUEST
12629
AATB12629EA
410
EUR
AATB12629
6XHis alkyne
6XHis alkyne
6XHis alkyne is an excellent building block to make 6XHis conjugates for developing His tag detection probes and purification tools through the well-known click chemistry.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only