IDA succinimidyl ester
Lieferant: AAT BIOQUEST
12632
AATB12632EA
429
EUR
AATB12632
IDA succinimidyl ester
IDA succinimidyl ester
IDA succinimidyl ester is an amino-reactive building block that can be used to develop a variety of IDA probes for detecting polyHis-containing proteins, and tools for purifying polyHis tagged proteins.
- Stable for 12 months
Immobilized metal affinity chromatography (IMAC) is a popular method for protein purification, particularly for recombinant proteins fused to a polyhistidine-tag.
Bitte beachten For Research Use Only