10XHis succinimidyl ester
Lieferant: AAT BIOQUEST
12640
AATB12640EA
549
EUR
AATB12640
10XHis succinimidyl ester
10XHis succinimidyl ester
10XHis succinimidyl ester is an excellent building block to make 10XHis conjugates for developing His tag detection probes and purification tools.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only