10XHis maleimide
Lieferant: AAT Bioquest
12641
AATB12641EA 549 EUR
AATB12641
10XHis maleimide
10XHis maleimide
10XHis maleimide is an excellent building block to make 10XHis conjugates for developing His tag detection probes and purification tools.
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only