HIS Lite™ Cy5 Bis NTA Chelator
Lieferant: AAT BIOQUEST
12653
AATB12653EA 619 EUR
AATB12653
HIS Lite™ Cy5 Bis NTA Chelator
Proteinkonjugate und Markierungen
Cy5 Bis NTA-Ni Chelator can be applied either to an SDS-PAGE gel or Western blot membrane for fluorescence imaging or used in living cells in combination with the addition of a heavy metal ion such as Ni²⁺.
  • Stable for 24 months
Quality tested in-house.
Bitte beachten For research use only.