HIS Lite™ OG488-Tris NTA Chelator
Lieferant: AAT BIOQUEST
12655
AATB12655EA 619 EUR
AATB12655
HIS Lite™ OG488-Tris NTA Chelator
Proteinkonjugate und Markierungen
HIS Lite™ OG488-Tris NTA Chelator is used as a sensitive green fluorescent probe for detecting polyhistidine-labelled proteins in combination with the addition of a heavy metal ion such as Ni²⁺ in cells, solution and solid surfaces.
  • Stable for 24 months
Quality tested in-house.
Bitte beachten For research use only.