HIS Lite™ iFluor® 647 Tris NTA Chelator
Lieferant: AAT BIOQUEST
12658
AATB12658EA
619
EUR
AATB12658
HIS Lite™ iFluor® 647 Tris NTA Chelator
Proteinkonjugate und Markierungen
iFluor® 647 Tris-NTA Chelator is used as a sensitive red fluorescent probe for detecting polyhistidine-labelled proteins in combination with the addition of a heavy metal ion such as Ni²⁺ in cells, solution and solid surfaces.
- Stable for 24 months
Quality tested in-house.
Bitte beachten For research use only.