HIS Lite™ Cy5 Tris NTA Chelator
Lieferant: AAT BIOQUEST
12659
AATB12659EA 619 EUR
AATB12659
HIS Lite™ Cy5 Tris NTA Chelator
Proteinkonjugate und Markierungen
Cy5-Tris NTA compound is a sensitive fluorescent chelator for detecting polyhistidine-labelled proteins in cells, solution and solid surfaces with the addition of certain metal ions (such as Ni²⁺, Co²⁺ etc.) as a mediator.
  • Stable for 24 months
Quality tested in-house.
Bitte beachten For research use only.