HIS Lite™ Cy5 Tris NTA Chelator
Lieferant: AAT BIOQUEST
Cy5-Tris NTA compound is a sensitive fluorescent chelator for detecting polyhistidine-labelled proteins in cells, solution and solid surfaces with the addition of certain metal ions (such as Ni²⁺, Co²⁺ etc.) as a mediator.
- Stable for 24 months
Quality tested in-house.
Bitte beachten For research use only.