HIS Lite™ Cy3 Tris NTA Chelator
Lieferant: AAT BIOQUEST
12660
AATB12660EA
619
EUR
AATB12660
HIS Lite™ Cy3 Tris NTA Chelator
Proteinkonjugate und Markierungen
Cy3-Tris NTA compound is a sensitive fluorescent chelator for detecting polyhistidine-labelled proteins in cells, solution, and solid surfaces with the addition of certain metal ions (such as Ni²⁺, Co₂⁺ and more) as a mediator.
- Stable for 24 months
Quality tested in-house.
Bitte beachten For research use only.