5-FITC cadaverine ≥95%
Lieferant: AAT BIOQUEST
Synonyme: 5-((5-Aminopentyl)thioureidyl)fluorescein
129
AATB129EA 460 EUR
AATB129
5-FITC cadaverine ≥95%
5-FITC cadaverine
FITC cadaverine is an excellent building block for developing fluoresceinated bioconjugates.
  • Multiple Bioconjugation Applications
  • Excellent Quality
Bitte beachten For research use only.