Fluorescein-5-maleimide ≥90%

Lieferant: AAT Bioquest
Achtung

130
AATB130EA 196 EUR
AATB130
Fluorescein-5-maleimide ≥90%
Fluorescein-5-maleimide

Maleimides are among the most frequently used reagents for thiol modification.

Formel: C₂₄H₁₃NO₇
MDL: MFCD00077345
CAS-Nummer: 75350-46-8
ADR: 5,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR