Benzoyl-Leu-Gly-Arg-Aminoluciferin
Lieferant: AAT BIOQUEST
Achtung
13000
AATB13000EA 769 EUR
AATB13000
Benzoyl-Leu-Gly-Arg-Aminoluciferin
Benzoyl-Leu-Gly-Arg-Aminoluciferin
This aminoluciferin-based probe is a luminogenic substrate for complement component C3/C5 convertases, coagulation factor Xa, and soybean trypsin-like enzyme.
  • High sensitivity
  • Excellent quality luminogenic substrate
  • Adaptable to multiple types of luciferase assays
ADR: 5,II