Benzoyl-Leu-Gly-Arg-Aminoluciferin
Lieferant: AAT BIOQUEST
|
Achtung
|
13000
AATB13000EA
769
EUR
AATB13000
Benzoyl-Leu-Gly-Arg-Aminoluciferin
Benzoyl-Leu-Gly-Arg-Aminoluciferin
This aminoluciferin-based probe is a luminogenic substrate for complement component C3/C5 convertases, coagulation factor Xa, and soybean trypsin-like enzyme.
- High sensitivity
- Excellent quality luminogenic substrate
- Adaptable to multiple types of luciferase assays
ADR:
5,II |