BOC-Val-Pro-Arg-Aminoluciferin
Lieferant: AAT BIOQUEST
Achtung
13002
AATB13002EA 769 EUR
AATB13002
BOC-Val-Pro-Arg-Aminoluciferin
BOC-Val-Pro-Arg-Aminoluciferin
BOC-Val-Pro-Arg-Aminoluciferin is a sentive luminescent substrate for alpha-thrombin.
  • High sensitivity
  • Excellent quality luminogenic substrate
  • Adaptable to multiple types of luciferase assays
ADR: 5 II