TF2-HQKLVFFAK-TQ2 [TF2-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Lys-TQ2]
Lieferant: AAT BIOQUEST
13206
AATB13206EA
965
EUR
AATB13206
TF2-HQKLVFFAK-TQ2 [TF2-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Lys-TQ2]
Proteine und Peptide
TF2-HQKLVFFAK-TQ2 [TF2-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Lys-TQ2] has much stronger fluoresce upon protease cleavage than the commonly used FRET peptide MCA-HQKLVFFA (K-DNP), a secretase FRET peptide substrate.
- Stable for 12 months
- Optimised for FRET applications
Quality tested in-house.
Bitte beachten For research use only.