Ac-DEVD-aminoluciferin
Lieferant: AAT BIOQUEST
Ac-DEVD-aminoluciferin is a luminogenic substrate that measures caspase-3 and -7 activities in apoptotic cells.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only