Ac-DEVD-aminoluciferin
Lieferant: AAT BIOQUEST
13210
AATB13210EA 133 EUR
AATB13210
Ac-DEVD-aminoluciferin
Ac-DEVD-aminoluciferin
Ac-DEVD-aminoluciferin is a luminogenic substrate that measures caspase-3 and -7 activities in apoptotic cells.
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only