Z-DEVD-aminoluciferin
Lieferant: AAT BIOQUEST
13211
AATB13211EA
271
EUR
AATB13211
Z-DEVD-aminoluciferin
Z-DEVD-aminoluciferin
Z-DEVD-aminoluciferin is a luminogenic substrate that measures caspase-3 and -7 activities in apoptotic cells.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only