Z-LETD-aminoluciferin
Lieferant: AAT BIOQUEST
13212
AATB13212EA 965 EUR
AATB13212
Z-LETD-aminoluciferin
Z-LETD-aminoluciferin
Z-LETD-aminoluciferin is a luminogenic substrate that measures caspase-8 activity in apoptotic cells.
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only