Z-LEHD-aminoluciferin
Lieferant: AAT BIOQUEST
13213
AATB13213EA 965 EUR
AATB13213
Z-LEHD-aminoluciferin
Z-LEHD-aminoluciferin
Z-LEHD-aminoluciferin is a luminogenic substrate that measures caspase-9 activity in apoptotic cells.
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only