Z-IETD-aminoluciferin
Lieferant: AAT BIOQUEST
13214
AATB13214EA 1250 EUR
AATB13214
Z-IETD-aminoluciferin
Z-IETD-aminoluciferin
Z-IETD-aminoluciferin is a luminogenic substrate that measures caspase-6 and granzyme B activities in apoptotic cells.
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only