Z-VDVAD-aminoluciferin
Lieferant: AAT BIOQUEST
13215
AATB13215EA
2020
EUR
AATB13215
Z-VDVAD-aminoluciferin
Z-VDVAD-aminoluciferin
Z-VDVAD-aminoluciferin is a luminogenic substrate that measures caspase-2 activity in apoptotic cells.
- Stable for 24 months
Quality tested in-house