Z-VEID-aminoluciferin
Lieferant: AAT BIOQUEST
13216
AATB13216EA
1250
EUR
AATB13216
Z-VEID-aminoluciferin
Z-VEID-aminoluciferin
Z-VEID-aminoluciferin is a luminogenic substrate that measures caspase-6 activity in apoptotic cells.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only