Z-ATAD-aminoluciferin
Lieferant: AAT BIOQUEST
13217
AATB13217EA 1250 EUR
AATB13217
Z-ATAD-aminoluciferin
Z-ATAD-aminoluciferin
Z-ATAD-aminoluciferin is a luminogenic substrate that measures caspase-12 activity in apoptotic cells.
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only