Z-VAD(OMe)-FMK
Lieferant: AAT BIOQUEST
13300
AATB13300EA 143 EUR
AATB13300
Z-VAD(OMe)-FMK
Proteine und Peptide
Z-VAD-FMK (carbobenzoxy-valyl-alanyl-aspartyl-[O-methyl]- fluoromethylketone) is a cell-permeable, irreversible pan caspase inhibitor that binds to the catalytic site of caspase proteases and can inhibit induction of apoptosis.
  • Caspase and apoptosis inhibitor
  • Enhanced stability
  • Increased cell permeability
For: Molecular biology, Labeling, Proteomics
GHS Symbol: GHS07
Hazard Class 5
Packing Group II
Signalword: Warning