Z-DEVD-FMK

Lieferant: AAT BIOQUEST
Achtung

13303
AATB13303EA 143 EUR
AATB13303
Z-DEVD-FMK
Z-DEVD-FMK

Z-DEVD-FMK is a specific, irreversible Caspase-3 inhibitor, and also shows potent inhibition on caspase-6, caspase-7, caspase-8, and caspase-10.


  • Multiple caspases inhibitor
  • Enhanced stability
  • Increased cell permeability

Formel: C₃₀H₄₁FN₄O₁₂
Molecular Weight: 668,66 g/mol
MDL: MFCD23380217
CAS-Nummer: 210344-95-9
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR