Z-LETD-FMK
Lieferant: AAT BIOQUEST
|
Achtung
|
13307
AATB13307EA
143
EUR
AATB13307
Z-LETD-FMK
Z-LETD-FMK
Z-LETD-FMK is a cell permeable, irreversible caspase protease 8 inhibitor that binds to the catalytic site of caspase- 8, which plays an important role in the induction of apoptosis.
- Caspase and apoptosis inhibitor
- Minimal cytotoxicity
- Cell permeable
ADR:
5,II |