Z-LETD-FMK
Lieferant: AAT BIOQUEST
Achtung
13307
AATB13307EA 143 EUR
AATB13307
Z-LETD-FMK
Z-LETD-FMK
Z-LETD-FMK is a cell permeable, irreversible caspase protease 8 inhibitor that binds to the catalytic site of caspase- 8, which plays an important role in the induction of apoptosis.
  • Caspase and apoptosis inhibitor
  • Minimal cytotoxicity
  • Cell permeable
ADR: 5,II