Ac-DEVD-AFC
Lieferant: AAT BIOQUEST
13401
AATB13401EA 152 EUR
AATB13401
Ac-DEVD-AFC
Proteine und Peptide
Ac-DEVD-AFC is a fluorogenic substrate for caspase 3, a protease that is rapidly activated when cells are exposed to apoptotic conditions and that cleaves poly(ADP-ribose) polymerase.
  • Hazard Class/ Packing Group: 5, II
  • GHS symbol: GHS07
  • Signal Word: Warning