Ac-DEVD-AFC
Lieferant: AAT BIOQUEST
13401
AATB13401EA
152
EUR
AATB13401
Ac-DEVD-AFC
Proteine und Peptide
Ac-DEVD-AFC is a fluorogenic substrate for caspase 3, a protease that is rapidly activated when cells are exposed to apoptotic conditions and that cleaves poly(ADP-ribose) polymerase.
- Hazard Class/ Packing Group: 5, II
- GHS symbol: GHS07
- Signal Word: Warning