Ac-DEVD-CHO
Lieferant: AAT BIOQUEST
13403
AATB13403EA
152
EUR
AATB13403
Ac-DEVD-CHO
Proteine und Peptide
Ac-DEVD-CHO is a reversible inhibitor of IL-1ß converting enzyme (ICE); inhibits poly(ADP-ribose) polymerase cleavage by apopain (caspase 3).
- Hazard Class/ Packing Group: 5, II
- GHS symbol: GHS07
- Signal Word: Warning