Ac-IETD-AMC

Lieferant: AAT BIOQUEST
Achtung

13411
AATB13411EA 307 EUR
AATB13411
Ac-IETD-AMC
Ac-IETD-AMC

Ac-IETD-AMC is a fluorogenic caspase-8/granzyme B substrate containing the acetyl (Ac) moiety.


  • Caspase detection
  • Bright signal

Formel: C31H41N5O12
CAS-Nummer: 348079-17-4
ADR: 5,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR