6-Amino-D-luciferin Assay-Referenzstandard

Lieferant: AAT BIOQUEST
Achtung

Synonyme: (S)-2-(6-Amino-2-benzothiazolyl)-2-thiazoline-4-carboxylic acid, (S)-2-(6-Amino-2-benzothiazolyl)-4,5-dihydro-4-thiazolecarboxylic acid, 6′-Amino-D-luciferin

13415
AATB13415EA 307 EUR
AATB13415
6-Amino-D-luciferin Assay-Referenzstandard
6-Amino-D-luciferin

D-aminoluciferin is a precursor that is widely used for developing luminescent protease substrates.


  • Subtrate precursor
  • Luminescence assay reference standard

Formel: C₁₁H₉N₃O₂S₂
Molecular Weight: 279,34 g/mol
CAS-Nummer: 161055-47-6
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3eb4fe506971c1ec21d940219303d5fae7c00f2e&vid=d4a3548b8acd5ab751cfc0bb5e0c4aa1b9014f14&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR