FAM-LETD-FMK
Lieferant: AAT BIOQUEST
Achtung
13416
AATB13416EA 383 EUR
AATB13416
FAM-LETD-FMK
FAM-LETD-FMK
Activation of caspases plays a central role in apoptosis.
  • Caspase detection
  • Minimal cytotoxicity
  • Cell permeable
ADR: 5,II