FITC-C6-D(OMe)E(OMe)VD(OMe)-FMK
Lieferant: AAT BIOQUEST
Achtung
13417 13418
AATB13417EA 307 EUR
AATB13417 AATB13418
FITC-C6-D(OMe)E(OMe)VD(OMe)-FMK
FITC-C6-D(OMe)E(OMe)VD(OMe)-FMK
Activation of caspases plays a central role in apoptosis.
  • Caspase detection
  • Minimal cytotoxicity
  • Cell permeable
ADR: 5 II