Cbz-IETD-AFC ≥90%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: Z-Ile-Glu-Thr-Asp-AFC

13425
AATB13425EA 196 EUR
AATB13425
Cbz-IETD-AFC ≥90%
Cbz-IETD-AFC

Z-IETD-AFC is a fluorogenic caspase-8/granzyme B substrate containing the acetyl (Ac) moiety.

Formel: C₃₇H₄₂F₃N₅O₁₃
MDL: MFCD01318832
CAS-Nummer: 219138-02-0
ADR: 5,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR