Ac-Leu-Glu-His-Asp-AMC ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
N-Acetyl-Leu-Glu-His-Asp-7-amino-4-methylcoumarin
13426
AATB13426EA
230
EUR
AATB13426
Ac-Leu-Glu-His-Asp-AMC ≥95%
Ac-Leu-Glu-His-Asp-AMC
Caspase 9 is a member of the CED-3 subfamily of the caspase family of cysteine proteases that play an essential role in the execution phase of apoptosis.
Formel:
C₃₃H₄₁N₇O₁₁ |
MDL:
MFCD03788104 CAS-Nummer: 292633-16-0 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...