(Ac-LEHD)2-R110
Lieferant: AAT BIOQUEST
|
Achtung
|
13427
AATB13427EA
152
EUR
AATB13427
(Ac-LEHD)2-R110
(Ac-LEHD)2-R110
Caspase 9 is a member of the CED-3 subfamily of the caspase family of cysteine proteases that play an essential role in the execution phase of apoptosis.
- Caspase detection
- Bright signal
ADR:
5,II |