(Ac-IETD)2-R110
Lieferant: AAT BIOQUEST
|
Achtung
|
13431
AATB13431EA
152
EUR
AATB13431
(Ac-IETD)2-R110
(Ac-IETD)2-R110
Since highly purified rhodamine 110 (R110)-derived substrates are locked in the lactone configuration they are colorless and non-fluorescent.
- Caspase detection
- Bright signal
ADR:
5,II |