(Ac-IETD)2-R110
Lieferant: AAT BIOQUEST
Achtung
13431
AATB13431EA 152 EUR
AATB13431
(Ac-IETD)2-R110
(Ac-IETD)2-R110
Since highly purified rhodamine 110 (R110)-derived substrates are locked in the lactone configuration they are colorless and non-fluorescent.
  • Caspase detection
  • Bright signal
ADR: 5,II