L-Leucine(4-methyl-7-coumarinylamide) hydrochloride ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: H-Leu-AMC·HCl

13445
AATB13445EA 230 EUR
AATB13445
L-Leucine(4-methyl-7-coumarinylamide) hydrochloride ≥95%
L-Leucine(4-methyl-7-coumarinylamide) hydrochloride

Leu-AMC is a fluorogenic substrate for leucine aminopeptidase.

Formel: C₁₆H₂₀N₂O₃·HCl
Molecular Weight: 324,805 g/mol
MDL: MFCD00043262
CAS-Nummer: 62480-44-8
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR