Ac-LKKR-AMC
Lieferant: AAT BIOQUEST
13446
AATB13446EA 277 EUR
AATB13446
Ac-LKKR-AMC
Ac-LKKR-AMC
Ac-Leu-Lys-Lys-Arg-AMC is a fluorogenic aminopeptidase and endopeptidase substrate.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only