Gly pro-7-amido-4-methylcoumarin hydrobromide ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
H-Gly-Pro-AMC·HBr
13450
AATB13450EA
152
EUR
AATB13450
Gly pro-7-amido-4-methylcoumarin hydrobromide ≥95%
Gly pro-7-amido-4-methylcoumarin hydrobromide
Gly-Pro-AMC is a sensitive fluorogenic substrate for detecting dipeptidyl peptidase IV (DPPIV), a serine protease that cleaves N-terminal dipeptides from polypeptides with L-proline or L-alanine at the penultimate position.
Formel:
C₁₇H₂₀BrN₃O₄ Molecular Weight: 410,268 g/mol |
MDL:
MFCD00152092 CAS-Nummer: 115035-46-6 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...