(Suc-LLVY)2R110
Lieferant: AAT BIOQUEST
|
Achtung
|
13451
AATB13451EA
152
EUR
AATB13451
(Suc-LLVY)2R110
(Suc-LLVY)2R110
(Suc-LLVY)2R110 is a sentive fluorogenic substrate for 20S proteasome, calpains and other chymotrypsin-like proteases.
- Protease substrate
- Bright signal
- Multiple applications
ADR:
5,II |