(Suc-LLVY)2R110
Lieferant: AAT BIOQUEST
Achtung
13451
AATB13451EA 152 EUR
AATB13451
(Suc-LLVY)2R110
(Suc-LLVY)2R110
(Suc-LLVY)2R110 is a sentive fluorogenic substrate for 20S proteasome, calpains and other chymotrypsin-like proteases.
  • Protease substrate
  • Bright signal
  • Multiple applications
ADR: 5,II