Suc-LLVY-Aminoluciferin
Lieferant: AAT BIOQUEST
|
Achtung
|
13452
AATB13452EA
307
EUR
AATB13452
Suc-LLVY-Aminoluciferin
Suc-LLVY-Aminoluciferin
Suc-LLVY-D-Aminolucifin is a sentive luminogenic substrate for 20S proteasome, calpains and other chymotrypsin-like proteases.
- Protease substrate
- Bright signal
- Multiple applications
ADR:
5,II |