Suc-LLVY-Aminoluciferin
Lieferant: AAT BIOQUEST
Achtung
13452
AATB13452EA 307 EUR
AATB13452
Suc-LLVY-Aminoluciferin
Suc-LLVY-Aminoluciferin
Suc-LLVY-D-Aminolucifin is a sentive luminogenic substrate for 20S proteasome, calpains and other chymotrypsin-like proteases.
  • Protease substrate
  • Bright signal
  • Multiple applications
ADR: 5,II